Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARO4
Entrez Gene ID
822269
Gene Name
armadillo repeat only 4
Gene Aliases
AT3G26600, armadillo repeat only 4
Chromosome Location
Chr.3: 9769634 - 9772122 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001338814.1 NP_001327909.1 2 - 2 2097 108
NM_113569.3 NP_189292.1 1 - 1 2308 108
GenBank mRNA X97616.1 - - 2163 108

View Full Product Details