Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSH2
Entrez Gene ID
821383
Gene Name
MUTS homolog 2
Gene Aliases
AT3G18524, ATMSH2, MUTS homolog 2
Chromosome Location
Chr.3: 6367941 - 6372685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113607.4 NP_566804.3 4 - 5 1486 85
GenBank mRNA AF002706.1 - 4 - 5 1368 85
AF026549.1 - 4 - 5 1348 85

View Full Product Details