Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DXR
Entrez Gene ID
836400
Gene Name
1-deoxy-D-xylulose 5-phosphate reductoisomerase
Gene Aliases
1-DEOXY-D-XYLULOSE 5-PHOSPHATE REDUCTOISOMERASE, 1-deoxy-D-xylulose 5-phosphate reductoisomerase, AT5G62790, MQB2.90, MQB2_90, PDE129, PIGMENT-DEFECTIVE EMBRYO 129
Chromosome Location
Chr.5: 25214125 - 25217469 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203671.1 NP_001190600.1 6 - 7 757 75
NM_125674.3 NP_201085.1 5 - 6 742 75
GenBank mRNA AF148852.1 - 5 - 6 650 75
AY045634.1 - 5 - 6 689 75
AY050802.1 - 5 - 6 688 75
AY054682.1 - 5 - 6 650 75
AY081453.1 - 5 - 6 565 75
AY084775.1 - 5 - 6 650 75
AY091405.1 - 5 - 6 565 75
AY098952.1 - 5 - 6 565 75

View Full Product Details