Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAC10
Entrez Gene ID
836408
Gene Name
RAC-like 10
Gene Aliases
ARABIDOPSIS THALIANA RAC-LIKE 10, ARAC10, AT5G62880, ATRAC10, ATROP11, MQB2.180, MQB2_180, RAC-like 10, RHO-RELATED PROTEIN FROM PLANTS 11, RHO-related protein from plants 11, ROP11
Chromosome Location
Chr.5: 25236945 - 25239262 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345564.1 NP_001331034.1 4 - 5 650 79
NM_125682.5 NP_201093.1 4 - 5 632 79
GenBank mRNA AF079485.1 - 4 - 5 388 79
AK176893.1 - 4 - 5 439 79
BT002997.1 - 4 - 5 434 79
BT004459.1 - 4 - 5 359 79

View Full Product Details