Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDH
Entrez Gene ID
836509
Gene Name
histidinol dehydrogenase
Gene Aliases
AT5G63890, ATHDH, HISN8, HISTIDINE BIOSYNTHESIS 8, MGI19.9, MGI19_9, histidinol dehydrogenase
Chromosome Location
Chr.5: 25565431 - 25568206 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125784.3 NP_568981.2 1 - 2 180 116
GenBank mRNA AB027709.1 - 1 - 2 127 116
AY039881.1 - 1 - 2 169 116
AY143900.1 - 1 - 2 78 116

View Full Product Details