Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABA4a
Entrez Gene ID
836652
Gene Name
RAB GTPase homolog A4A
Gene Aliases
AT5G65270, AtRABA4a, MQN23.22, MQN23_22, RAB GTPase homolog A4A
Chromosome Location
Chr.5: 26083216 - 26084832 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125925.4 NP_201330.1 1 - 2 450 74
GenBank mRNA AY045947.1 - 1 - 2 290 74
AY079309.1 - 1 - 2 229 74
AY087004.1 - 1 - 2 290 74

View Full Product Details