Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COAB
Entrez Gene ID
837789
Gene Name
4-phospho-panto-thenoylcysteine synthetase
Gene Aliases
4-phospho-panto-thenoylcysteine synthetase, AT1G12350, ATCOAB, F5O11.36, F5O11_36
Chromosome Location
Chr.1: 4198337 - 4200849 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332031.1 NP_001321808.1 2 - 3 557 97
NM_101107.4 NP_563904.2 2 - 3 361 97
GenBank mRNA AK118086.1 - 2 - 3 285 97

View Full Product Details