Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AHP2
Entrez Gene ID
837893
Gene Name
Tat-binding protein 1(Tbp-1)-interacting protein (TBPIP)
Gene Aliases
AT1G13330, Arabidopsis Hop2 homolog, HOMOLOGOUS-PAIRING PROTEIN 2, HOP2, T6J4.9, T6J4_9
Chromosome Location
Chr.1: 4567935 - 4570598 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101204.3 NP_172791.1 6 - 7 767 100
GenBank mRNA AK176610.1 - 6 - 7 767 100
AK227903.1 - 6 - 7 767 100

View Full Product Details