The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_101210.3 | NP_172797.2 | 3 - 4 | 438 | 78 |
GenBank mRNA | AY465923.1 | - | 3 - 4 | 378 | 78 |