Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUB
Entrez Gene ID
837900
Gene Name
C2H2 and C2HC zinc fingers superfamily protein
Gene Aliases
AT1G13400, JAGGED-LIKE, JGL, NUBBIN, T6J4.23, T6J4_23
Chromosome Location
Chr.1: 4597567 - 4599141 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101210.3 NP_172797.2 4 - 5 526 106
GenBank mRNA AY465923.1 - 4 - 5 466 106

View Full Product Details