Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTR3
Entrez Gene ID
837998
Gene Name
UDP-galactose transporter 3
Gene Aliases
AT1G14360, ATUTR3, F14L17.13, F14L17_13, UDP-galactose transporter 3
Chromosome Location
Chr.1: 4911093 - 4913167 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_101303.4 NP_563949.1 4 - 5 499 95
GenBank mRNA AF332450.1 - 4 - 5 361 95
AY059942.1 - 4 - 5 484 95
AY114599.1 - 4 - 5 361 95

View Full Product Details