Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK30
Entrez Gene ID
843813
Gene Name
calcium-dependent protein kinase 30
Gene Aliases
AT1G74740, ATCPK30, CALCIUM-DEPENDENT PROTEIN KINASE 1A, CDPK1A, F25A4.29, F25A4_29, calcium-dependent protein kinase 30
Chromosome Location
Chr.1: 28079946 - 28082671 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106132.4 NP_177612.2 4 - 5 1248 127
GenBank mRNA AY140016.1 - 4 - 5 1221 127
BT008900.1 - 4 - 5 1053 127

View Full Product Details