Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH10A8
Entrez Gene ID
843831
Gene Name
aldehyde dehydrogenase 10A8
Gene Aliases
AT1G74920, F25A4.11, F25A4_11, aldehyde dehydrogenase 10A8
Chromosome Location
Chr.1: 28138997 - 28142837 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198470.1 NP_001185399.1 13 - 14 1468 81
NM_106150.4 NP_565094.1 13 - 14 1576 81
GenBank mRNA AY087395.1 - 13 - 14 1483 81
AY093071.1 - 13 - 14 1441 81
BT008872.1 - 13 - 14 1312 81

View Full Product Details