Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH10A8
Entrez Gene ID
843831
Gene Name
aldehyde dehydrogenase 10A8
Gene Aliases
AT1G74920, F25A4.11, F25A4_11, aldehyde dehydrogenase 10A8
Chromosome Location
Chr.1: 28138997 - 28142837 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198470.1 NP_001185399.1 7 - 8 921 74
NM_106150.4 NP_565094.1 7 - 8 1029 74
GenBank mRNA AY087395.1 - 7 - 8 936 74
AY093071.1 - 7 - 8 894 74
BT008872.1 - 7 - 8 765 74

View Full Product Details