Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXL4
Entrez Gene ID
843924
Gene Name
extracellular lipase 4
Gene Aliases
AT1G75910, T4O12.250, T4O12_250, extracellular lipase 4
Chromosome Location
Chr.1: 28501434 - 28503283 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106241.3 NP_177719.1 3 - 4 679 100
GenBank mRNA AK227028.1 - 3 - 4 677 100

View Full Product Details