Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMY2
Entrez Gene ID
843945
Gene Name
alpha-amylase-like 2
Gene Aliases
ARABIDOPSIS THALIANA ALPHA-AMYLASE-LIKE 2, AT1G76130, ATAMY2, T23E18.6, T23E18_6, alpha-amylase-like 2
Chromosome Location
Chr.1: 28560629 - 28564159 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334710.1 NP_001323338.1 7 - 8 1038 75
NM_106262.4 NP_177740.1 7 - 8 673 75
GenBank mRNA AK221564.1 - 7 - 8 673 75
AY084871.1 - 7 - 8 669 75
BT025560.1 - 6 - 7 586 75

View Full Product Details