Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CUC3
- Entrez Gene ID
-
843975
- Gene Name
- NAC (No Apical Meristem) domain transcriptional regulator superfamily protein
- Gene Aliases
-
ANAC031, AT1G76420, Arabidopsis NAC domain containing protein 31, CUP SHAPED COTYLEDON3, F15M4.8, NAC368
- Chromosome Location
-
Chr.1: 28671806 - 28674045 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106292.3 |
NP_177768.1 |
1 - 2 |
411 |
84 |
View Full Product Details