Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BGAL16
Entrez Gene ID
844078
Gene Name
beta-galactosidase 16
Gene Aliases
AT1G77410, F2P24.12, F2P24_12, beta-galactosidase 16
Chromosome Location
Chr.1: 29088657 - 29093264 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334779.1 NP_001320598.1 13 - 14 1831 80
NM_001334780.1 NP_001320597.1 14 - 15 1753 80
NM_001334781.1 NP_001320596.1 11 - 12 1327 80
NM_106391.2 NP_177866.2 14 - 15 1738 80

View Full Product Details