Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAX3
Entrez Gene ID
844105
Gene Name
like AUX1 3
Gene Aliases
AT1G77690, T32E8.2, T32E8_2, like AUX1 3
Chromosome Location
Chr.1: 29200417 - 29203418 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106418.4 NP_177892.1 3 - 4 567 123
GenBank mRNA AY054266.1 - 3 - 4 489 123
BT008300.1 - 3 - 4 466 123

View Full Product Details