Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PGM
- Entrez Gene ID
-
844140
- Gene Name
- phosphoglycerate/bisphosphoglycerate mutase
- Gene Aliases
-
AT1G78050, F28K19.30, F28K19_30, PHOSPHOGLYCERATE/BISPHOSPHOGLYCERATE MUTASE, phosphoglycerate/bisphosphoglycerate mutase
- Chromosome Location
-
Chr.1: 29347581 - 29349755 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106454.4 |
NP_177928.2 |
2 - 3 |
618 |
107 |
View Full Product Details