Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LZF1
Entrez Gene ID
844196
Gene Name
light-regulated zinc finger protein 1
Gene Aliases
AT1G78600, B-box domain protein 22, BBX22, DBB3, DOUBLE B-BOX 3, SALT TOLERANCE HOMOLOG 3, STH3, T30F21.7, T30F21_7, light-regulated zinc finger protein 1
Chromosome Location
Chr.1: 29566863 - 29568931 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106507.4 NP_565183.1 2 - 3 1139 89
GenBank mRNA AY087387.1 - 2 - 3 785 89
AY093108.1 - 2 - 3 791 89
BT006324.1 - 2 - 3 632 89

View Full Product Details