Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MSL6
- Entrez Gene ID
-
844197
- Gene Name
- mechanosensitive channel of small conductance-like 6
- Gene Aliases
-
AT1G78610, T30F21.6, T30F21_6, mechanosensitive channel of small conductance-like 6
- Chromosome Location
-
Chr.1: 29568961 - 29572572 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106508.3 |
NP_177982.1 |
3 - 4 |
2507 |
105 |
View Full Product Details