Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALIS5
Entrez Gene ID
844283
Gene Name
ALA-interacting subunit 5
Gene Aliases
ALA-interacting subunit 5, AT1G79450, T8K14.13, T8K14_13
Chromosome Location
Chr.1: 29887616 - 29890211 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106593.4 NP_565210.1 6 - 7 923 76
NM_202446.1 NP_974175.1 5 - 6 951 76
GenBank mRNA AY088777.1 - 6 - 7 804 76

View Full Product Details