Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ELF7
- Entrez Gene ID
-
844312
- Gene Name
- hydroxyproline-rich glycoprotein family protein
- Gene Aliases
-
AT1G79730, EARLY FLOWERING 7, F19K16.29, F19K16_29
- Chromosome Location
-
Chr.1: 30000538 - 30004083 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106622.5 |
NP_178091.2 |
6 - 7 |
1271 |
72 |
View Full Product Details