Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RABE1e
Entrez Gene ID
820148
Gene Name
RAB GTPase homolog E1E
Gene Aliases
AT3G09900, ATRAB8E, ATRABE1E, RAB GTPase homolog E1E
Chromosome Location
Chr.3: 3034477 - 3036961 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111825.4 NP_187601.1 3 - 4 452 142
GenBank mRNA AK229294.1 - 3 - 4 371 142
AY084345.1 - 3 - 4 284 142
BT024925.1 - 3 - 4 242 142

View Full Product Details