Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIP5K9
Entrez Gene ID
820151
Gene Name
phosphatidyl inositol monophosphate 5 kinase
Gene Aliases
AT3G09920, phosphatidyl inositol monophosphate 5 kinase
Chromosome Location
Chr.3: 3038780 - 3044989 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035589.3 NP_001030666.1 7 - 8 2530 67
NM_001202923.1 NP_001189852.1 7 - 8 2460 67
NM_001337850.1 NP_001327647.1 7 - 8 2474 67
NM_111827.3 NP_187603.1 6 - 7 3326 67
GenBank mRNA AJ810853.1 - 6 - 7 2060 67
AK229815.1 - 7 - 8 2309 67

View Full Product Details