Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUM4
Entrez Gene ID
820198
Gene Name
pumilio 4
Gene Aliases
APUM4, AT3G10360, pumilio 4
Chromosome Location
Chr.3: 3210884 - 3215621 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111871.7 NP_187647.2 8 - 9 2951 150

View Full Product Details