Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC8
Entrez Gene ID
820201
Gene Name
subunit of exocyst complex 8
Gene Aliases
AT3G10380, ATSEC8, SUBUNIT OF EXOCYST COMPLEX 8, subunit of exocyst complex 8
Chromosome Location
Chr.3: 3219507 - 3229034 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111873.4 NP_566372.1 26 - 27 3425 70
GenBank mRNA AK118541.1 - 26 - 27 3249 70
AY059763.1 - 26 - 27 3249 70

View Full Product Details