Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXO70E1
Entrez Gene ID
822601
Gene Name
exocyst subunit exo70 family protein E1
Gene Aliases
AT3G29400, ATEXO70E1, exocyst subunit exo70 family protein E1
Chromosome Location
Chr.3: 11297180 - 11299695 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113866.3 NP_189586.1 1 - 1 2222 69
GenBank mRNA BT008695.1 - 1 - 1 2148 69

View Full Product Details