Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COBL2
Entrez Gene ID
822697
Gene Name
COBRA-like protein 2 precursor
Gene Aliases
AT3G29810, COBRA-like protein 2 precursor
Chromosome Location
Chr.3: 11728024 - 11730350 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113910.5 NP_566851.1 3 - 4 800 72

View Full Product Details