Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BR6OX2
Entrez Gene ID
822709
Gene Name
brassinosteroid-6-oxidase 2
Gene Aliases
AT3G30180, CYP85A2, brassinosteroid-6-oxidase 2
Chromosome Location
Chr.3: 11810726 - 11813883 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113917.4 NP_566852.1 7 - 8 1320 153
GenBank mRNA AB087801.1 - 7 - 8 1218 153
AY052655.1 - 7 - 8 1270 153
AY063728.1 - 7 - 8 1179 153

View Full Product Details