Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC16
Entrez Gene ID
823831
Gene Name
plastid transcriptionally active 16
Gene Aliases
AT3G46780, plastid transcriptionally active 16
Chromosome Location
Chr.3: 17228642 - 17231229 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114545.3 NP_566886.2 4 - 5 1139 85

View Full Product Details