Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSA2
Entrez Gene ID
824034
Gene Name
glutamate-1-semialdehyde 2,1-aminomutase 2
Gene Aliases
1-aminomutase 2, AT3G48730, glutamate-1-semialdehyde 2
Chromosome Location
Chr.3: 18049571 - 18051778 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114732.5 NP_190442.1 1 - 2 302 92
GenBank mRNA AK229328.1 - 1 - 2 250 92
BT025324.1 - 1 - 2 176 92

View Full Product Details