Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DMS3
Entrez Gene ID
824086
Gene Name
defective in meristem silencing 3
Gene Aliases
AT3G49250, DEFECTIVE IN MERISTEM SILENCING 3, IDN1, INVOLVED IN DE NOVO 1
Chromosome Location
Chr.3: 18255960 - 18260998 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114784.3 NP_566916.1 6 - 7 1437 81
GenBank mRNA AY049288.1 - 6 - 7 1385 81

View Full Product Details