Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PEN3
- Entrez Gene ID
-
824173
- Gene Name
- PTEN 3
- Gene Aliases
-
AT3G50110, ATPEN3, Arabidopsis thaliana phosphatase and TENsin homolog deleted on chromosome ten 3, PTEN 3, PTEN2B, phosphatase and TENsin homolog deleted on chromosome ten 2B
- Chromosome Location
-
Chr.3: 18580609 - 18584813 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114871.3 |
NP_566929.1 |
11 - 12 |
1844 |
93 |
GenBank mRNA |
BT000931.1 |
- |
10 - 11 |
1382 |
93 |
View Full Product Details