Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KICP-02
Entrez Gene ID
824186
Gene Name
ATP binding microtubule motor family protein
Gene Aliases
AT3G50240
Chromosome Location
Chr.3: 18623254 - 18628914 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339461.1 NP_001326509.1 11 - 12 1599 84
NM_001339462.1 NP_001326510.1 10 - 11 1641 84
NM_114884.5 NP_566931.1 11 - 12 1599 84
GenBank mRNA AB061676.1 - 11 - 12 1574 84

View Full Product Details