Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KICP-02
Entrez Gene ID
824186
Gene Name
ATP binding microtubule motor family protein
Gene Aliases
AT3G50240
Chromosome Location
Chr.3: 18623254 - 18628914 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339461.1 NP_001326509.1 15 - 16 2114 95
NM_001339462.1 NP_001326510.1 14 - 15 2156 95
NM_001339463.1 NP_001326508.1 15 - 16 2099 95
NM_114884.5 NP_566931.1 15 - 16 2114 95
GenBank mRNA AB061676.1 - 15 - 16 2089 95

View Full Product Details