Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSBO2
Entrez Gene ID
824246
Gene Name
photosystem II subunit O-2
Gene Aliases
AT3G50820, MSP-2, OEC33, OXYGEN EVOLVING COMPLEX SUBUNIT 33 KDA, PHOTOSYSTEM II SUBUNIT O-2, PSBO, PSBO-2, photosystem II subunit O-2
Chromosome Location
Chr.3: 18890874 - 18892508 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114942.3 NP_190651.1 1 - 2 437 86
GenBank mRNA AJ145957.1 - 1 - 2 276 86
AY050362.1 - 1 - 2 298 86
AY088799.1 - 1 - 2 355 86
AY116934.1 - 1 - 2 240 86

View Full Product Details