Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GASA2
Entrez Gene ID
826546
Gene Name
GAST1 protein homolog 2
Gene Aliases
AT4G09610, GAST1 PROTEIN HOMOLOG, GAST1 protein homolog 2, T25P22.50, T25P22_50
Chromosome Location
Chr.4: 6074770 - 6075645 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117029.2 NP_192699.1 1 - 2 131 90
GenBank mRNA U11765.1 - 1 - 2 111 90

View Full Product Details