Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYTC-2
Entrez Gene ID
826595
Gene Name
cytochrome c-2
Gene Aliases
AT4G10040, F28M11.5, cytochrome c-2
Chromosome Location
Chr.4: 6276948 - 6278622 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117072.4 NP_192742.1 1 - 2 251 71
GenBank mRNA AY045944.1 - 1 - 2 201 71
AY079373.1 - 1 - 2 116 71
AY087056.1 - 1 - 2 194 71

View Full Product Details