Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYTC-2
Entrez Gene ID
826595
Gene Name
cytochrome c-2
Gene Aliases
AT4G10040, F28M11.5, cytochrome c-2
Chromosome Location
Chr.4: 6276948 - 6278622 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117072.4 NP_192742.1 2 - 3 386 102
GenBank mRNA AY045944.1 - 2 - 3 336 102
AY079373.1 - 2 - 3 251 102
AY087056.1 - 2 - 3 329 102

View Full Product Details