Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX6
Entrez Gene ID
826765
Gene Name
glutathione peroxidase 6
Gene Aliases
AT4G11600, ATGPX6, LSC803, PHGPX, T5C23.30, T5C23_30, glutathione peroxidase 6
Chromosome Location
Chr.4: 7009769 - 7011406 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117229.4 NP_192897.2 3 - 4 452 62
GenBank mRNA AY039863.1 - 3 - 4 401 62
AY077655.1 - 3 - 4 376 62

View Full Product Details