Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDB1
Entrez Gene ID
828937
Gene Name
NAD(P)H dehydrogenase B1
Gene Aliases
AT4G28220, F26K10.100, F26K10_100, NAD(P)H dehydrogenase B1
Chromosome Location
Chr.4: 13992210 - 13995831 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118962.5 NP_567801.1 7 - 8 2380 73
GenBank mRNA BT025339.1 - 7 - 8 1512 73

View Full Product Details