Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CKL6
Entrez Gene ID
828972
Gene Name
casein kinase I-like 6
Gene Aliases
AT4G28540, F20O9.240, F20O9_240, PAPK1, casein kinase I-like 6
Chromosome Location
Chr.4: 14106735 - 14110706 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118996.5 NP_567812.1 9 - 10 1277 145
GenBank mRNA AY099715.1 - 9 - 10 1017 145
AY943845.1 - 9 - 10 728 145
BT000286.1 - 9 - 10 728 145

View Full Product Details