Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDX19
Entrez Gene ID
832129
Gene Name
nudix hydrolase homolog 19
Gene Aliases
ARABIDOPSIS THALIANA NUDIX HYDROLASE HOMOLOG 19, AT5G20070, ATNUDT19, ATNUDX19, F28I16.220, F28I16_220, nudix hydrolase homolog 19
Chromosome Location
Chr.5: 6779841 - 6782501 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122014.4 NP_197507.1 5 - 6 1083 73
GenBank mRNA AK226922.1 - 5 - 6 1047 73
AY049280.1 - 5 - 6 1064 73
AY090287.1 - 5 - 6 1031 73

View Full Product Details