Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XIK
Entrez Gene ID
832171
Gene Name
myosin family protein with Dil
Gene Aliases
AT5G20490, ATXIK, F7C8.80, F7C8_80, MYOSIN XI K, MYOSIN XI-17, XI-17, XI-K
Chromosome Location
Chr.5: 6926516 - 6937830 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161252.2 NP_001154724.1 19 - 20 2330 67
NM_001343670.1 NP_001318611.1 19 - 20 2309 67
NM_001343671.1 NP_001318612.1 20 - 21 2762 67
GenBank mRNA HQ427882.1 - 19 - 20 2312 67

View Full Product Details