Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGH
Entrez Gene ID
832372
Gene Name
SWAP (Suppressor-of-White-APricot)/surp domain-containing protein
Gene Aliases
AT5G23080, MYJ24.7, MYJ24_7, TOUGH
Chromosome Location
Chr.5: 7742940 - 7749020 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122214.4 NP_197699.2 9 - 10 993 110
GenBank mRNA AK118129.1 - 9 - 10 901 110
BT008588.1 - 9 - 10 862 110

View Full Product Details