Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGA3
Entrez Gene ID
838812
Gene Name
transcription factor TGA3
Gene Aliases
AT1G22070, F2E2.14, F2E2_14, TGA1A-related gene 3, TRANSCRIPTION FACTOR TGA3
Chromosome Location
Chr.1: 7789133 - 7792139 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102057.4 NP_564156.1 3 - 4 813 64
GenBank mRNA BT026034.1 - 2 - 3 295 64
L10209.1 - 3 - 4 376 64

View Full Product Details