Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUM8
Entrez Gene ID
838829
Gene Name
pumilio 8
Gene Aliases
APUM8, AT1G22240, F16L1.3, F16L1_3, pumilio 8
Chromosome Location
Chr.1: 7852904 - 7855213 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102074.2 NP_173643.1 1 - 2 1040 91

View Full Product Details