Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LHT2
- Entrez Gene ID
-
839057
- Gene Name
- lysine histidine transporter 2
- Gene Aliases
-
AATL2, AMINO ACID TRANSPORTER-LIKE PROTEIN 2, ARABIDOPSIS LYSINE HISTIDINE TRANSPORTER 2, AT1G24400, ATLHT2, F21J9.6, lysine histidine transporter 2
- Chromosome Location
-
Chr.1: 8651407 - 8653915 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102286.3 |
NP_564217.1 |
4 - 5 |
1421 |
62 |
GenBank mRNA |
AB046210.1 |
- |
4 - 5 |
1088 |
62 |
View Full Product Details